Thermo Scientific Chemicals

4-Nitrophenylhydrazine mono and dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L09273.04
2 g, Each
Thermo Scientific Chemicals

4-Nitrophenylhydrazine mono and dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: L09273.04
2 g, Each
Quantity
Catalog number: L09273.04
also known as L09273-04
Price (USD)
Have Questions?
Assay (HPLC)
≥97.5%
Appearance (Color)
Pink to orange to brown
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder

Description

4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water, DMSO and methanol.

Notes
Hygroscopic. Incompatible materials are Strong oxidizing agents and moisture. Store under dry inert gas. Protect from humidity nd water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text