1-Adamantaneethanol, 98%, Thermo Scientific Chemicals
1-Adamantaneethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Adamantaneethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L09318.06
also known as L09318-06
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
1-Adamantaneethanol, 98%, Thermo Scientific Chemicals
Catalog numberL09318.06
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS6240-11-5
IUPAC Name2-(adamantan-1-yl)ethan-1-ol
Molecular FormulaC12H20O
InChI KeyZBIDZPHRNBZTLT-UHFFFAOYSA-N
SMILESOCCC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Appearance (Color)White
Formcrystalline powder
Melting Point70-78?C (≤3?C range)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only