Thermo Scientific Chemicals

Isopropyl acetate, 99+%, Thermo Scientific Chemicals

Catalog number: L09337.AK
250 mL, Each
Thermo Scientific Chemicals

Isopropyl acetate, 99+%, Thermo Scientific Chemicals

Catalog number: L09337.AK
250 mL, Each
Quantity
Catalog number: L09337.AK
also known as L09337-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-21-4
IUPAC Name
propan-2-yl acetate
Molecular Formula
C5H10O2
InChI Key
JMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILES
CC(C)OC(C)=O
Assay (GC)
≥99.0% (UK-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Refractive Index
1.3755-1.3785 @ 20?C (UK-sourced material)
Appearance (Color)
Clear colorless
Transition Temperature
≥99.0% (US-sourced material)

Description

Isopropyl acetate is a solvent in chemical industry, especially for cellulose, plastics, waxes, resins, gums, paints, oil and fats. and also as flavoring agent. It is an active component of perfumes and printing inks. It is also employed as an extractant for the preparation of antibiotics, vitamins and hormones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl acetate is a solvent in chemical industry, especially for cellulose, plastics, waxes, resins, gums, paints, oil and fats. and also as flavoring agent. It is an active component of perfumes and printing inks. It is also employed as an extractant for the preparation of antibiotics, vitamins and hormones.

Solubility
Slightly miscible with cold water and methanol.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text