Thermo Scientific Chemicals

2-Naphthylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L09384.14
25 g, Each
Thermo Scientific Chemicals

2-Naphthylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L09384.14
25 g, Each
Quantity
Catalog number: L09384.14
also known as L09384-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7498-57-9
IUPAC Name
2-(naphthalen-2-yl)acetonitrile
Molecular Formula
C12H9N
InChI Key
LPCWDVLDJVZIHA-UHFFFAOYSA-N
SMILES
N#CCC1=CC=C2C=CC=CC2=C1
Melting Point (clear melt)
81.5-87.5?C
Appearance (Color)
White to cream to yellow
Form
Powder
Assay (GC)
≥96.0%

Description

2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text