1-Tetradecanethiol, 94%, Thermo Scientific Chemicals
1-Tetradecanethiol, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Tetradecanethiol, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09401.06
also known as L09401-06
Price (USD)/ Each
38.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
38.70
Add to cart
1-Tetradecanethiol, 94%, Thermo Scientific Chemicals
Catalog numberL09401.06
Price (USD)/ Each
38.70
-
Add to cart
Chemical Identifiers
CAS2079-95-0
IUPAC Nametetradecane-1-thiol
Molecular FormulaC14H30S
InChI KeyGEKDEMKPCKTKEC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥92.0%
Refractive Index1.4575-1.4645 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only