Thermo Scientific Chemicals

Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L09416.03
1 g, Each
Thermo Scientific Chemicals

Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L09416.03
1 g, Each
Quantity
Catalog number: L09416.03
also known as L09416-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5239-82-7
IUPAC Name
N-(1,3-dimethyl-1,3,2-diazaborolidin-2-yl)-S-[(1,3-dimethyl-1,3,2-diazaborolidin-2-yl)imino]-λ⁴-sulfanimine
Molecular Formula
C8H20B2N6S
InChI Key
BXXOHRWKRXBZTG-UHFFFAOYSA-N
SMILES
CN1CCN(C)B1N=S=NB1N(C)CCN1C
Appearance (Color)
Clear colorless
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Liquid or viscous liquid
Refractive Index
1.4325-1.4385 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text