Thermo Scientific Chemicals

(S)-(+)-3-Methyl-2-butanol, 99%, Thermo Scientific Chemicals

Catalog number: L09431.ME
500 mg, Each
Thermo Scientific Chemicals

(S)-(+)-3-Methyl-2-butanol, 99%, Thermo Scientific Chemicals

Catalog number: L09431.ME
500 mg, Each
Quantity
Catalog number: L09431.ME
also known as L09431-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1517-66-4
IUPAC Name
3-methylbutan-2-ol
Molecular Formula
C5H12O
InChI Key
MXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILES
CC(C)C(C)O
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4080-1.4110 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

It is an pharmaceutical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pharmaceutical and organic intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text