Thermo Scientific Chemicals

4-Amino-3-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog number: L09487.14
25 g, Each
Thermo Scientific Chemicals

4-Amino-3-nitrophenol, 98%, Thermo Scientific Chemicals

Catalog number: L09487.14
25 g, Each
Quantity
Catalog number: L09487.14
also known as L09487-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-81-1
Form
Crystalline powder or powder
Appearance (Color)
Dark red to brown to dark brown
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
145-155?C

Description

4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-nitrophenol was used in the synthesis of benzimidazole based Factor Xa (trypsin-like serine protease) inhibitors and 4-(4-amino-3-nitrophenoxy)phthalonitrile. It was also employed as matrix during visible matrix-assisted laser desorption/ionization mass spectrometry.

Solubility
Soluble in water. (1.8 g/L.)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text