Thermo Scientific Chemicals

Hexaketocyclohexane octahydrate, 99%, Thermo Scientific Chemicals

Catalog number: L09488.09
10 g, Each
Thermo Scientific Chemicals

Hexaketocyclohexane octahydrate, 99%, Thermo Scientific Chemicals

Catalog number: L09488.09
10 g, Each
Quantity
Catalog number: L09488.09
also known as L09488-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7255-28-9
Appearance (Color)
White to cream to brown
Assay (unspecified)
≥98.5% (by C elemental assay)
Elemental Analysis
C: 22.74-23.43% (Theory: 23.08%)
Form
Powder

Description

Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaketocyclohexane octahydrate is used in the preparation of hexaazatriphenylenehexacarbonitrile. It is used as pharmaceutical intermediates.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text