Thermo Scientific Chemicals

2-n-Butoxyethyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: L09515.0F
2500 mL, Each
Thermo Scientific Chemicals

2-n-Butoxyethyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: L09515.0F
2500 mL, Each
Quantity
Catalog number: L09515.0F
also known as L09515-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-07-2
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4115-1.4155 @ 20?C
Identification (FTIR)
Conforms

Description

2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

Solubility
Miscible with hydrocarbons and organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text