2-n-Butoxyethyl acetate, 98%
2-n-Butoxyethyl acetate, 98%
2-n-Butoxyethyl acetate, 98%
2-n-Butoxyethyl acetate, 98%
Thermo Scientific Chemicals

2-n-Butoxyethyl acetate, 98%

CAS: 112-07-2 | C8H16O3 | 160.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number L09515.AP
also known as L09515-AP
Price (USD)
42.80
Each
In stock
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
42.80
Each
Add to cart
Chemical Identifiers
CAS112-07-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4115-1.4155 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Ask our AI about this Product
2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

Solubility
Miscible with hydrocarbons and organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Magami, S. M.; Oldring, P. K. T.; Castle, L.; Guthrie, J. T. The physical-chemical behaviour of amino cross-linkers and the effect of their chemistry on selected epoxy can coatings’ hydrolysis to melamine and to formaldehyde into aqueous food simulants. Prog. Org. Coat. 2015, 78, 325-333.
  2. Magami, S. M.; Oldring, P. K. T.; Castle, L.; Guthrie, J. T. The effect of TiO2, pigmentation on the hydrolysis of amino resin crosslinked epoxy can coatings. J. Coat. Technol. Res. 2014, 11 (6), 1013-1022.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
10238931Certificate of AnalysisFeb 20, 2024L09515.AP, L09515.AE, L09515.0F
10249208Certificate of AnalysisDec 29, 2023L09515.AP, L09515.AE, L09515.0F
10246354Certificate of AnalysisMay 09, 2023L09515.0F
10218561Certificate of AnalysisFeb 15, 2023L09515.AE, L09515.0F, L09515.AP, ALF-L09515-0F, ALF-L09515-AE, ALF-L09515-AP
10188171Certificate of AnalysisDec 08, 2021L09515.AP, L09515.AE, L09515.0F
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link