Thermo Scientific Chemicals

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%, Thermo Scientific Chemicals

Catalog number: L09516.06
5 g, Each
Thermo Scientific Chemicals

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%, Thermo Scientific Chemicals

Catalog number: L09516.06
5 g, Each
Quantity
Catalog number: L09516.06
also known as L09516-06
Price (USD)
Quantity
-
Have Questions?

Description

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text