3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals
3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L09518.09
also known as L09518-09
Price (USD)/ Each
71.70
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
71.70
Add to cart
3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals
Catalog numberL09518.09
Price (USD)/ Each
71.70
-
Add to cart
Chemical Identifiers
CAS4439-02-5
IUPAC Name2-(2H-1,3-benzodioxol-5-yl)acetonitrile
Molecular FormulaC9H7NO2
InChI KeyZQPBOYASBNAXOZ-UHFFFAOYSA-N
SMILESN#CCC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Assay (GC)≥98.0%
FormCrystals or crystalline powder and/or fused solid
Melting Point (clear melt)39-45?C
3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chia-lin J. Wang.; William C. Ripka.; Pat N. Confalone. A short and stereospecific synthesis of (±)-α-lycorane .Tetrahedron Lett. 1984, 25 (41),4613-4616 .
  2. Winston A. Davis.; Michael P. Cava. A new synthesis of arylmalononitriles.J. Org. Chem. 1983, 48 (16),2774-2775 .