Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: L09518.18
50 g, Each
Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: L09518.18
50 g, Each
Quantity
Catalog number: L09518.18
also known as L09518-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4439-02-5
IUPAC Name
2-(2H-1,3-benzodioxol-5-yl)acetonitrile
Molecular Formula
C9H7NO2
InChI Key
ZQPBOYASBNAXOZ-UHFFFAOYSA-N
SMILES
N#CCC1=CC=C2OCOC2=C1
Appearance (Color)
White to cream to yellow
Assay (GC)
≥98.0%
Form
Crystals or crystalline powder and/or fused solid
Melting Point (clear melt)
39-45?C

Description

3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text