It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.
Solubility
Insoluble in water. Soluble in DMSO.
Notes
It may be stored in the dark in a desiccator. Avoid contact with strong heat and metal salts.
RUO – Research Use Only
Santosh D. Diwakara.; Sachin S. Bhagwat.; Murlidhar S. Shingare.; Charansing H. Gill. Substituted 3-((Z)-2-(4-nitrophenyl)-2-(1H-tetrazol-5-yl) vinyl)-4H-chromen-4-ones as novel anti-MRSA agents: Synthesis, SAR, and in-vitro assessment.Bioorg. Med. Chem. Lett. 2008, 18 (16),4678-4681 .