Thermo Scientific Chemicals

2,2,3,4,4,4-Hexafluorobutyl methacrylate, 96%, stab., Thermo Scientific Chemicals

Catalog number: L09543.14
25 g, Each
Thermo Scientific Chemicals

2,2,3,4,4,4-Hexafluorobutyl methacrylate, 96%, stab., Thermo Scientific Chemicals

Catalog number: L09543.14
25 g, Each
Quantity
Catalog number: L09543.14
also known as L09543-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36405-47-7
IUPAC Name
(3S)-2,2,3,4,4,4-hexafluorobutyl 2-methylprop-2-enoate
Molecular Formula
C8H8F6O2
InChI Key
DFVPUWGVOPDJTC-LURJTMIESA-N
SMILES
CC(=C)C(=O)OCC(F)(F)[C@H](F)C(F)(F)F
Appearance (Color)
Clear, colorless
Assay (GC)
>95.0%
Solution Test
10% w/v (in Diethyl ether): clear
Form
Liquid
Refractive Index
1.3590-1.3650 @ 20?C

Description

It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text