Thermo Scientific Chemicals

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%, Thermo Scientific Chemicals

Catalog number: L09558.14
25 g, Each
Thermo Scientific Chemicals

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%, Thermo Scientific Chemicals

Catalog number: L09558.14
25 g, Each
Quantity
Catalog number: L09558.14
also known as L09558-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35132-20-8
IUPAC Name
(1R,2R)-1,2-diphenylethane-1,2-bis(aminium)
Molecular Formula
C14H18N2
InChI Key
PONXTPCRRASWKW-ZIAGYGMSSA-P
SMILES
[NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat, air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text