2,4-Hexadiyn-1,6-diol, 97%, Thermo Scientific Chemicals
2,4-Hexadiyn-1,6-diol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Hexadiyn-1,6-diol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09576.03
also known as L09576-03
Price (USD)/ Each
63.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
63.80
Add to cart
2,4-Hexadiyn-1,6-diol, 97%, Thermo Scientific Chemicals
Catalog numberL09576.03
Price (USD)/ Each
63.80
-
Add to cart
Chemical Identifiers
CAS3031-68-3
IUPAC Namehexa-2,4-diyne-1,6-diol
Molecular FormulaC6H6O2
InChI KeyJXMQYKBAZRDVTC-UHFFFAOYSA-N
SMILESOCC#CC#CCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (GC)>96.0%
Appearance (Color)Cream to brown or pale orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only