Thermo Scientific Chemicals

4,4'-Diaminodicyclohexylmethane, mixture of stereoisomers, 98+%, Thermo Scientific Chemicals

Catalog number: L09598.22
100 g, Each
Thermo Scientific Chemicals

4,4'-Diaminodicyclohexylmethane, mixture of stereoisomers, 98+%, Thermo Scientific Chemicals

Catalog number: L09598.22
100 g, Each
Quantity
Catalog number: L09598.22
also known as L09598-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1761-71-3
IUPAC Name
4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Molecular Formula
C13H26N2
InChI Key
DZIHTWJGPDVSGE-UHFFFAOYSA-N
SMILES
NC1CCC(CC2CCC(N)CC2)CC1
Assay (GC)
≥98.0% (sum of stereoisomers)
Form
Fused solid or clear liquid as melt
Appearance (Color)
White or colorless to pale brown
Identification (FTIR)
Conforms

Description

4,4'-Diaminodicyclohexylmethane is used as epoxy curing agents. It is a versatile intermediate to produce leather, rubber products, plastics, dyes, and photo sensitive polymers. It is used in manufacturing diisocyanates and polyamides. It is also used as lubricant additive as and corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminodicyclohexylmethane is used as epoxy curing agents. It is a versatile intermediate to produce leather, rubber products, plastics, dyes, and photo sensitive polymers. It is used in manufacturing diisocyanates and polyamides. It is also used as lubricant additive as and corrosion inhibitor.

Solubility
Soluble in water. (5 g/L) at 20°C.

Notes
Incompatible materials are acids, acid chlorides, acid anhydrides, oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text