Thermo Scientific Chemicals

Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific Chemicals

Catalog number: L09616.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 3-isocyanatopropionate, 98%, Thermo Scientific Chemicals

Catalog number: L09616.03
1 g, Each
Quantity
Catalog number: L09616.03
also known as L09616-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5100-34-5
IUPAC Name
ethyl 3-isocyanatopropanoate
Molecular Formula
C6H9NO3
InChI Key
XBSGYVHOINMTIM-UHFFFAOYSA-N
SMILES
CCOC(=O)CCN=C=O
Refractive Index
1.4260-1.4330 @ 20°C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colourless
Form
Liquid

Description

Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.

Solubility
Not miscible with water.

Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text