Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
Solubility
Not miscible with water.
Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.
RUO – Research Use Only