Thermo Scientific Chemicals

1-Hexanethiol, 96%, Thermo Scientific Chemicals

Catalog number: L09659.AE
100 mL, Each
Thermo Scientific Chemicals

1-Hexanethiol, 96%, Thermo Scientific Chemicals

Catalog number: L09659.AE
100 mL, Each
Quantity
Catalog number: L09659.AE
also known as L09659-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-31-9
IUPAC Name
hexane-1-thiol
Molecular Formula
C6H14S
InChI Key
PMBXCGGQNSVESQ-UHFFFAOYSA-N
SMILES
CCCCCCS
Refractive Index
1.4460-1.4520 @ 20°C (non-U.S. specification)
Form
Liquid
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥95.0%

Description

It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text