Thermo Scientific™

D-Phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog number: L09697.03
1 g, Each
Thermo Scientific™

D-Phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog number: L09697.03
1 g, Each
Quantity
Catalog number: L09697.03
also known as L09697-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5267-64-1
IUPAC Name
2-amino-3-phenylpropan-1-ol
Molecular Formula
C9H13NO
InChI Key
STVVMTBJNDTZBF-UHFFFAOYNA-N
SMILES
NC(CO)CC1=CC=CC=C1
Appearance (Color)
White to pale yellow
Assay (Non-aqueous acid-base Titration)
>97.5%
Form
crystalline powder
Optical Rotation
+23 ? 2? (c=5 in ethanol)

Description

D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

Solubility
Soluble in Dichloromethane, Ethyl Acetate, Methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with carbondioxide, air, acids and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text