Thermo Scientific Chemicals

3,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L09707.03
1 g, Each
Thermo Scientific Chemicals

3,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L09707.03
1 g, Each
Quantity
Catalog number: L09707.03
also known as L09707-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42288-46-0
IUPAC Name
2-(3,5-dimethylphenyl)acetate
Molecular Formula
C10H11O2
InChI Key
HDNBKTWQBJJYPD-UHFFFAOYSA-M
SMILES
CC1=CC(CC([O-])=O)=CC(C)=C1
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White to cream
Assay (Silylated GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
98.5-104.5?C

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text