3,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific Chemicals
3,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L09707.06
also known as L09707-06
Price (USD)/ Each
187.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
187.00
Add to cart
3,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific Chemicals
Catalog numberL09707.06
Price (USD)/ Each
187.00
-
Add to cart
Chemical Identifiers
CAS42288-46-0
IUPAC Name2-(3,5-dimethylphenyl)acetate
Molecular FormulaC10H11O2
InChI KeyHDNBKTWQBJJYPD-UHFFFAOYSA-M
SMILESCC1=CC(CC([O-])=O)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White to cream
Assay (Silylated GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)98.5-104.5?C
It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Henry Gilman.; Henry A. Pacevitz.; Ogden Baine. Benzylalkali Compounds.J. Am. Chem. Soc. 1940, 62 (6), 1514-1520 .
  2. John J. Bost.; Richard E. Kepner.; A. Dinsmoor Webb. Effect of Structural Changes on Adsorption of Certain Alcohol 3,5-dinitrobenzoates on Silicic Acid1. J. Org. Chem.. 1957, 22 (1),51-55 DOI: 10.1021/jo01352a013 .