n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Hossain, M. Z.; Teja, A. S. Modeling phase equilibria in CO 2+ polymer systems. J. Supercrit. Fluids 2015, 96, 313-323.
- Koç, D. İ.; Koç, M. L. A genetic programming-based QSPR model for predicting solubility parameters of polymers. Chemometr. Intell. Lab Syst. 2015, 144, 122-127.