Thermo Scientific Chemicals

Tetramethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L09716.14
25 g, Each
Thermo Scientific Chemicals

Tetramethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L09716.14
25 g, Each
Quantity
Catalog number: L09716.14
also known as L09716-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
681-84-5
IUPAC Name
tetramethyl silicate
Molecular Formula
C4H12O4Si
InChI Key
LFQCEHFDDXELDD-UHFFFAOYSA-N
SMILES
CO[Si](OC)(OC)OC
Assay (GC)
≥97.5% (UK Sourced Material)
Identification (FTIR)
Conforms (UK Sourced Material)
Form
Liquid
Comment
Material Sourced in UK and US
Appearance (Color)
Clear colorless

Description

Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

Solubility
Miscible with ethanol and organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text