trans-2-Butenyl acetate, 95%, Thermo Scientific Chemicals
trans-2-Butenyl acetate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Butenyl acetate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09734.06
also known as L09734-06
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
trans-2-Butenyl acetate, 95%, Thermo Scientific Chemicals
Catalog numberL09734.06
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS628-08-0
IUPAC Namebut-2-en-1-yl acetate
Molecular FormulaC6H10O2
InChI KeyWNHXJHGRIHUOTG-UHFFFAOYSA-N
SMILESCC=CCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4135-1.4210 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only