Thermo Scientific Chemicals

3,5-Dimethylbenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L09748.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dimethylbenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L09748.06
5 g, Each
Quantity
Catalog number: L09748.06
also known as L09748-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5779-95-3
IUPAC Name
3,5-dimethylbenzaldehyde
Molecular Formula
C9H10O
InChI Key
NBEFMISJJNGCIZ-UHFFFAOYSA-N
SMILES
CC1=CC(C=O)=CC(C)=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.5365-1.5405 @ 20?C
Form
Liquid
Free acid (titration)
≤1.5%

Description

3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

Solubility
Not miscible with water.

Notes
Handle and store under inert gas. Air Sensitive. Store away from air and oxidizing agents. Incompatible with oxidizing agents and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text