Thermo Scientific™

1,4-Phenylene diisothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L09756.03
1 g, Each
Thermo Scientific™

1,4-Phenylene diisothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L09756.03
1 g, Each
Quantity
Catalog number: L09756.03
also known as L09756-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4044-65-9
IUPAC Name
1,4-diisothiocyanatobenzene
Molecular Formula
C8H4N2S2
InChI Key
OMWQUXGVXQELIX-UHFFFAOYSA-N
SMILES
S=C=NC1=CC=C(C=C1)N=C=S
Melting Point (clear melt)
127.0-134.0°C
Assay from Supplier's CofA
≥97.5% (U.S. specification)
Appearance (Color)
White to cream to yellow to brown to gray
Assay (GC)
≥97.5% (non-U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)

Description

It is used in the treatment of hookworms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the treatment of hookworms.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text