1,2-Dimethylnaphthalene, 98%, Thermo Scientific Chemicals
1,2-Dimethylnaphthalene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dimethylnaphthalene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L09774.MD
also known as L09774-MD
Price (USD)/ Each
27.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
27.80
Add to cart
1,2-Dimethylnaphthalene, 98%, Thermo Scientific Chemicals
Catalog numberL09774.MD
Price (USD)/ Each
27.80
-
Add to cart
Chemical Identifiers
CAS573-98-8
IUPAC Name1,2-dimethylnaphthalene
Molecular FormulaC12H12
InChI KeyQNLZIZAQLLYXTC-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=CC2=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay from Supplier's CofA≥97.5%
FormLiquid
1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
RUO – Research Use Only

General References:

  1. Milton L. Lee.; Daniel L. Vassilaros.; Curt M. White. Retention indices for programmed-temperature capillary-column gas chromatography of polycyclic aromatic hydrocarbons.Anal. Chem. 1979, 51 (6),768-773 .
  2. Arthur E. Jacobson.; Everette L. May. Structures Related to Morphine. XXXI.1 2'-Substituted Benzomorphans. J. Med. Chem. 1965, 8 (5),563-566 .