Thermo Scientific Chemicals

2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: L09783.09
10 g, Each
Thermo Scientific Chemicals

2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals

Catalog number: L09783.09
10 g, Each
Quantity
Catalog number: L09783.09
also known as L09783-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1888-94-4
Form
Liquid
Assay from Suppliers CofA
≥96.0%
Appearance (Color)
Clear colorless to yellow
Stabilizer
400-600ppm hydroquinone

Description

2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text