2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals
2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L09783.18
also known as L09783-18
Price (USD)/ Each
367.65
Online exclusive
408.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
367.65
Online exclusive
408.00 
Save 40.35 (10%)
Add to cart
2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals
Catalog numberL09783.18
Price (USD)/ Each
367.65
Online exclusive
408.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1888-94-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofA≥96.0%
Appearance (Color)Clear colorless to yellow
Stabilizer400-600ppm hydroquinone
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. He, X.; Liu, Z.; Fan, F.; Qiang, S.; Cheng, L.; Yang, W. Poly(ionic liquids) hollow nanospheres with PDMAEMA as joint support of highly dispersed gold nanoparticles for thermally adjustable catalysis. J. Nanopart. Res. 2015, 17 (2), 1-10.
  2. Rajalakshmi, K.; Krishnan, P. S. G.; Nayak, S. K. Synthesis of dialkyl 2-(Methacryloyloxyethyl) phosphonates, their characterization and polymerization. Polym. Sci. Ser. B 2015, 57 (5), 408-416.