Thermo Scientific Chemicals

2-Chlorophenylacetone, 96%, Thermo Scientific Chemicals

Catalog number: L09846.03
1 g, Each
Thermo Scientific Chemicals

2-Chlorophenylacetone, 96%, Thermo Scientific Chemicals

Catalog number: L09846.03
1 g, Each
Quantity
Catalog number: L09846.03
also known as L09846-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6305-95-9
IUPAC Name
1-(2-chlorophenyl)propan-2-one
Molecular Formula
C9H9ClO
InChI Key
LWGNDIMNCPMZOF-UHFFFAOYSA-N
SMILES
CC(=O)CC1=CC=CC=C1Cl
Form
Liquid
Refractive Index
1.5325-1.5385 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥95.0%

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text