Thermo Scientific Chemicals

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%, Thermo Scientific Chemicals

Catalog number: L09855.06
5 g, Each
Thermo Scientific Chemicals

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, 99%, Thermo Scientific Chemicals

Catalog number: L09855.06
5 g, Each
Quantity
Catalog number: L09855.06
also known as L09855-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22204-53-1
IUPAC Name
2-(6-methoxynaphthalen-2-yl)propanoic acid
Molecular Formula
C14H14O3
InChI Key
CMWTZPSULFXXJA-UHFFFAOYNA-N
SMILES
COC1=CC=C2C=C(C=CC2=C1)C(C)C(O)=O
Melting Point (clear melt)
153-158?C
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
+66.0 ? 2.0? (c=1 in chloroform)
Assay (Silylated GC)
≥98.5%

Description

(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid is a non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text