Thermo Scientific Chemicals

Dibenzyl phosphite, 90+%, Thermo Scientific Chemicals

Catalog number: L09858.06
5 g, Each
Thermo Scientific Chemicals

Dibenzyl phosphite, 90+%, Thermo Scientific Chemicals

Catalog number: L09858.06
5 g, Each
Quantity
Catalog number: L09858.06
also known as L09858-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17176-77-1
IUPAC Name
bis(benzyloxy)(oxo)-λ⁵-phosphanylium
Molecular Formula
C14H14O3P
InChI Key
RQKYHDHLEMEVDR-UHFFFAOYSA-N
SMILES
O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Assay by NMR
≥90%
Refractive Index
1.5500-1.5580 @ 20?C
Form
Liquid

Description

Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.

Solubility
Soluble in DMSO, Methanol. Reacts with water.

Notes
Store at 4°C. Moisture Sensitive. Incompatible with moisture and oxidizing agents. Store under dry inert gas. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text