DL-Arabinose, 98+%
Thermo Scientific Chemicals

DL-Arabinose, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number L09895.18
also known as L09895-18
Price (USD)/ Each
316.65
Online exclusive
352.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
316.65
Online exclusive
352.00 
Save 35.35 (10%)
Add to cart
DL-Arabinose, 98+%
Catalog numberL09895.18
Price (USD)/ Each
316.65
Online exclusive
352.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS147-81-9
IUPAC Nameoxane-2,3,4,5-tetrol
Molecular FormulaC5H10O5
InChI KeySRBFZHDQGSBBOR-UHFFFAOYNA-N
SMILESOC1COC(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Optical Rotation0? ? 10.0? (c=10, water + NH₄OH)
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%
DL-Arabinose is used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars. DL-Arabinose, is a building block of polysaccharide (CCPa-1) from fungus Coprinus comatus Gray, which has potential application as natural antitumor agent with immunomodulatory activity. Arabinose is also used as a culture medium for certain bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Arabinose is used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars. DL-Arabinose, is a building block of polysaccharide (CCPa-1) from fungus Coprinus comatus Gray, which has potential application as natural antitumor agent with immunomodulatory activity. Arabinose is also used as a culture medium for certain bacteria.

Solubility
Soluble in Water: 250 mg/5ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A G M Leijdekkers; J P M Bink; S Geutjes; H A Schols; H Gruppen. Enzymatic saccharification of sugar beet pulp for the production of galacturonic acid and arabinose; a study on the impact of the formation of recalcitrant oligosaccharides.Bioresource Technology.2013, 128 518-525.
  2. Hugo Mélida; Jose V Sandoval-Sierra; Javier Diéguez-Uribeondo; Vincent Bulone. Analyses of extracellular carbohydrates in oomycetes unveil the existence of three different cell wall types.Eukaryotic Cell.2013, 12 (2), 194-203