2-Chloro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
2-Chloro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L09899.06
also known as L09899-06
Price (USD)/ Each
67.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
67.10
Add to cart
2-Chloro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
Catalog numberL09899.06
Price (USD)/ Each
67.10
-
Add to cart
Chemical Identifiers
CAS50528-86-4
IUPAC Name1-chloro-2-isocyanato-4-(trifluoromethyl)benzene
Molecular FormulaC8H3ClF3NO
InChI KeyWEPYOPYMWSHRIW-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Cl)C(=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4880-1.4930 @ 20?C
Appearance (Color)Clear colorless to white
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only