Thermo Scientific Chemicals

Diethyl chlorophosphite, 97%, Thermo Scientific Chemicals

Catalog number: L09919.14
25 g, Each
Thermo Scientific Chemicals

Diethyl chlorophosphite, 97%, Thermo Scientific Chemicals

Catalog number: L09919.14
25 g, Each
Quantity
Catalog number: L09919.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
589-57-1
IUPAC Name
diethyl chlorophosphonite
Molecular Formula
C4H10ClO2P
InChI Key
TXHWYSOQHNMOOU-UHFFFAOYSA-N
SMILES
CCOP(Cl)OCC
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Form
Liquid
Refractive Index
1.4355-1.4395 @ 20?C

Description

Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.

Solubility
Soluble in DMSO and chloroform. Reacts with water.

Notes
Air & Moisture Sensitive. Store away from air and strong bases. Incompatible with water, bases, active metals and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text