Thermo Scientific Chemicals

Thiazole, 99%, Thermo Scientific Chemicals

Catalog number: L09970.14
25 g, Each
Thermo Scientific Chemicals

Thiazole, 99%, Thermo Scientific Chemicals

Catalog number: L09970.14
25 g, Each
Quantity
Catalog number: L09970.14
also known as L09970-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
288-47-1
IUPAC Name
1,3-thiazole
Molecular Formula
C3H3NS
InChI Key
FZWLAAWBMGSTSO-UHFFFAOYSA-N
SMILES
S1C=CN=C1
Form
Liquid
Refractive Index
1.5330-1.5395 @ 20°C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

Thiazole is used as a flavoring agent and in the preparation of dyes and rubber accelerators. It serves as a component of the vitamin thiamine (B1). It acts as a protected formyl group used in natural product synthesis. It reacts with alkyl lithium and Grignard’s reagent to prepare organometallic complexes. It is involved in the electrophilic aromatic substitution and nucleophilic aromatic substitution at C-5 and C-2 positions respectively. Further, it undergoes alkylation reaction to get thiazolium cation, which is used as a catalyst in the Stetter reaction and the Benzoin condensation. In addition to this, it is involved in the preparation of alagebrium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole is used as a flavoring agent and in the preparation of dyes and rubber accelerators. It serves as a component of the vitamin thiamine (B1). It acts as a protected formyl group used in natural product synthesis. It reacts with alkyl lithium and Grignard’s reagent to prepare organometallic complexes. It is involved in the electrophilic aromatic substitution and nucleophilic aromatic substitution at C-5 and C-2 positions respectively. Further, it undergoes alkylation reaction to get thiazolium cation, which is used as a catalyst in the Stetter reaction and the Benzoin condensation. In addition to this, it is involved in the preparation of alagebrium.

Solubility
Soluble in ethanol and ether. Slightly soluble in water.

Notes
Air and light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text