Thermo Scientific Chemicals

Sodium 3-nitrobenzoate, 94%, may contain up to ca 10% water, Thermo Scientific Chemicals

Catalog number: L10028.22
100 g, Each
Thermo Scientific Chemicals

Sodium 3-nitrobenzoate, 94%, may contain up to ca 10% water, Thermo Scientific Chemicals

Catalog number: L10028.22
100 g, Each
Quantity
Catalog number: L10028.22
also known as L10028-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
827-95-2
IUPAC Name
sodium 3-nitrobenzoate
Molecular Formula
C7H4NNaO4
InChI Key
MUADFEZFSKAZLT-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C1=CC=CC(=C1)[N+]([O-])=O
Form
Powder
Water Content (Karl Fischer Titration)
≤10.0%
Appearance (Color)
White to pale yellow or pale brown
Assay (HPLC)
>92.5%

Description

Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text