Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 3-nitrobenzoate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
RUO – Research Use Only
Zhiqiang Hu; Richard A Ferraina; Jon F Ericson; Allison A Mackay; Barth F Smets. Biomass characteristics in three sequencing batch reactors treating a wastewater containing synthetic organic chemicals.Water Research.2005, 39 (4), 710-720.
L J Nadeau; J C Spain. Bacterial degradation of m-nitrobenzoic acid.Applied and Environmental Microbiology.1995, 61 (2), 840-843.