Methyl 2-acetamidoacrylate, 97%, Thermo Scientific Chemicals
Methyl 2-acetamidoacrylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-acetamidoacrylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L10049.06
also known as L10049-06
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
Add to cart
Methyl 2-acetamidoacrylate, 97%, Thermo Scientific Chemicals
Catalog numberL10049.06
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS35356-70-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)48-53?C
Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kotha, S.; Bandarugattu, V. B.; Krishna, N. G. Diversity-oriented approach to unusual amino acid derivatives and heterocycles via methyl 2-acetamidoacrylate and its congeners. Tetrahedron 2014, 70 (35), 5361-5384.
  2. Basauri-Molina, M.; Riemersma, C. F.; Würdemann, M. A.; Kleijn, H.; Gebbink, R. K. Lipase active site covalent anchoring of Rh (NHC) catalysts: towards chemoselective artificial metalloenzymes. Chem. Commun. 2015, 51 (31), 6792-6795.