Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Hiroshi Tsubomura.; Robert P. Lang. Molecular Complexes and Their Spectra. XIII. Complexes of Iodine with Amides, Diethyl Sulfide and Diethyl Disulfide.J. Am. Chem. Soc. 1961, 83 (9),2085-2092 .