(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals
(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L10069.03
also known as L10069-03
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
161.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
161.00 
Save 24.35 (15%)
Add to cart
(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals
Catalog numberL10069.03
Price (USD)/ Each
136.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
161.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
CAS513-49-5
IUPAC Namebutan-2-amine
Molecular FormulaC4H11N
InChI KeyBHRZNVHARXXAHW-UHFFFAOYNA-N
SMILESCCC(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Optical Rotation8.2 ± 0.3? (neat)
Assay (GC)≥97.5%
Refractive Index1.3910-1.3950 @ 20?C
View more
It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert Oberlender.; Robert C. Pfaff.; Michael P. Johnson.; Xuemei Huang.; David E. Nichols. Stereoselective LSD-like activity in d-lysergic acid amides of R- and S-2-aminobutane.J. Med. Chem. 1992, 35 (2),203-211 .
  2. Harlan L. Goering.; Jon N. Eikenberry.; Gerald S. Koermer. Tris[3-(trifluoromethylhydroxymethylene)-d-camphorato]europium(III). Chiral shift reagent for direct determination of enantiomeric compositions.J. Am. Chem. Soc. 1971, 93 (22), 5913-5914.