Thermo Scientific Chemicals

(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals

Catalog number: L10069.06
5 g, Each
Thermo Scientific Chemicals

(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals

Catalog number: L10069.06
5 g, Each
Quantity
Catalog number: L10069.06
also known as L10069-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
513-49-5
IUPAC Name
butan-2-amine
Molecular Formula
C4H11N
InChI Key
BHRZNVHARXXAHW-UHFFFAOYNA-N
SMILES
CCC(C)N
Form
Liquid
Appearance (Color)
Clear colorless
Optical Rotation
8.2 ± 0.3? (neat)
Assay (GC)
≥97.5%
Refractive Index
1.3910-1.3950 @ 20?C

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text