Thermo Scientific Chemicals

4-Nitrophenyl-alpha-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: L10070.06
5 g, Each
Thermo Scientific Chemicals

4-Nitrophenyl-alpha-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

Catalog number: L10070.06
5 g, Each
Quantity
Catalog number: L10070.06
also known as L10070-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3767-28-0
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Formula
C12H15NO8
InChI Key
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
SMILES
OC[C@H]1O[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
Assay from Supplier's CofA
≥98.0% (US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)
Optical Rotation
+216? ±6? (c=1 in Water, US-sourced material)
Appearance (Color)
White to pale cream
Form
Crystalline powder

Description

4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

Solubility
Soluble in water, warm ethanol and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text