Thermo Scientific Chemicals

Methylhydrazine sulfate, 98%, Thermo Scientific Chemicals

Catalog number: L10122.14
25 g, Each
Thermo Scientific Chemicals

Methylhydrazine sulfate, 98%, Thermo Scientific Chemicals

Catalog number: L10122.14
25 g, Each
Quantity
Catalog number: L10122.14
also known as L10122-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
302-15-8
IUPAC Name
2-methylhydrazinium hydrogen sulfate
Molecular Formula
CH8N2O4S
InChI Key
KJDJPXUIZYHXEZ-UHFFFAOYSA-N
SMILES
CN[NH3+].OS([O-])(=O)=O
Appearance (Color)
White to pale cream to pale yellow
Form
Crystals or powder or crystalline powder and/or chunks
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
140.0-148.0°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text