Thermo Scientific Chemicals

Glycidyl 2-methylphenyl ether, tech. 85%, Thermo Scientific Chemicals

Catalog number: L10136.22
100 g, Each
Thermo Scientific Chemicals

Glycidyl 2-methylphenyl ether, tech. 85%, Thermo Scientific Chemicals

Catalog number: L10136.22
100 g, Each
Quantity
Catalog number: L10136.22
also known as L10136-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2210-79-9
IUPAC Name
2-[(2-methylphenoxy)methyl]oxirane
Molecular Formula
C10H12O2
InChI Key
KFUSXMDYOPXKKT-UHFFFAOYNA-N
SMILES
CC1=CC=CC=C1OCC1CO1
Appearance (Color)
Clear colorless to very pale yellow
Comment
Material Sourced in UK and US
Assay (GC)
≥80.0%
Refractive Index
1.5250-1.5330 @20?C (UK Sourced Material)
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text