Thermo Scientific Chemicals

Benzofuroxan, 98%, Thermo Scientific Chemicals

Catalog number: L10144.18
50 g, Each
Thermo Scientific Chemicals

Benzofuroxan, 98%, Thermo Scientific Chemicals

Catalog number: L10144.18
50 g, Each
Quantity
Catalog number: L10144.18
also known as L10144-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
480-96-6
IUPAC Name
2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Formula
C6H4N2O2
InChI Key
OKEAMBAZBICIFP-UHFFFAOYSA-N
SMILES
[O-][N+]1=C2C=CC=CC2=NO1
Appearance (Color)
Yellow to brown
Melting Point (clear melt)
65-71?C
Assay (GC)
≥97.5%
Form
Powder and/or lumpy solid

Description

Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text